Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290707.1 | NP_001277636.1 | 3 | 564 | ||
NM_011866.2 | NP_035996.2 | 2 | 518 | |||
GenBank mRNA | AF110507.1 | - | 2 | 389 | ||
AK014090.1 | - | 2 | 571 | |||
AK039249.1 | - | 3 | 564 | |||
AK046535.1 | - | 4 | 739 | |||
AK147690.1 | - | 2 | 485 | |||
AK162221.1 | - | 2 | 172 | |||
AY360383.1 | - | 2 | 518 | |||
BC113201.1 | - | 2 | 384 | 40048238 |