Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011790.4 | NP_035920.1 | 3, 4 | 565 | ||
GenBank mRNA | AF124664.1 | - | 2, 3 | 392 | ||
AJ130975.1 | - | 2, 3 | 424 | |||
AK011094.1 | - | 3, 4 | 569 | |||
AK045012.1 | - | 548 | ||||
AK149936.1 | - | 3, 4 | 545 | |||
AK165028.1 | - | 3, 4 | 503 | |||
AK167104.1 | - | 3, 4 | 522 | |||
AK168055.1 | - | 3, 4 | 570 | |||
BC051998.1 | - | 3, 4 | 565 | 6400287 | ||
BC052422.1 | - | 3, 4 | 565 | 6400287 |