Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011830.3 | NP_035960.2 | 4 | 300 | ||
GenBank mRNA | AK002676.1 | - | 4 | 332 | ||
AK011777.1 | - | 4 | 333 | |||
AK143505.1 | - | 4 | 341 | |||
AK146362.1 | - | 4 | 366 | |||
AK150713.1 | - | 4 | 336 | |||
AK151233.1 | - | 4 | 336 | |||
AK151706.1 | - | 4 | 359 | |||
AK152691.1 | - | 4 | 333 | |||
AK159822.1 | - | 4 | 333 | |||
AK160569.1 | - | 4 | 333 | |||
AK166667.1 | - | 4 | 335 | |||
AK169129.1 | - | 4 | 366 | |||
AK169788.1 | - | 4 | 366 | |||
AK215538.1 | - | 2 | 113 | |||
BC010314.1 | - | 4 | 300 | 2646679 | ||
BC052671.1 | - | 4 | 321 | 30058399 | ||
M33934.1 | - | 4 | 333 | |||
M98333.1 | - | 4 | 267 |