Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011872.2 | NP_036002.1 | 3 | 232 | ||
GenBank mRNA | AB008371.1 | - | 3 | 209 | ||
AF124299.1 | - | 3 | 234 | |||
AK029477.1 | - | 3 | 232 | |||
AK077406.1 | - | 3 | 232 | |||
BC027823.1 | - | 3 | 393 | 5322691 |