Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011809.3 | NP_035939.3 | 10 | 2408 | ||
GenBank mRNA | AK014602.2 | - | 10 | 2330 | ||
AK030537.1 | - | 6 | 1710 | |||
AK038001.1 | - | 1 | 2169 | |||
AK038348.1 | - | 10 | 2395 | |||
AK128933.1 | - | 11 | 2282 | |||
AK143687.1 | - | 10 | 2360 | |||
AK144974.1 | - | 10 | 2359 | |||
AK145852.1 | - | 10 | 2244 | |||
AK151315.1 | - | 10 | 2359 | |||
AK152750.1 | - | 10 | 2362 | |||
AK153478.1 | - | 10 | 2360 | |||
BC005486.1 | - | 10 | 2329 | 3486021 | ||
BC005504.1 | - | 10 | 2269 | 3490813 |