Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp54a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011899.4 | NP_036029.2 | 2, 3 | 359 | ||
GenBank mRNA | AK011928.1 | - | 2, 3 | 323 | ||
BC019683.1 | - | 2, 3 | 288 | 3708250 |
Gene Symbol: Srp54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100109.1 | NP_001093579.1 | 2, 3 | 595 | ||
GenBank mRNA | AK010716.1 | - | 2, 3 | 295 | ||
AK136033.1 | - | 2, 3 | 596 | |||
X16319.1 | - | 324 |
Gene Symbol: Srp54c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100110.1 | NP_001093580.1 | 285 | |||
GenBank mRNA | AK090018.1 | - | 288 | |||
AK146826.1 | - | 323 | ||||
BC005543.1 | - | 302 | 3585216 |