Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011906.2 | NP_036036.2 | 5 | 715 | ||
GenBank mRNA | AF051098.1 | - | 5 | 522 | ||
AK089317.1 | - | 5 | 586 | |||
AK155116.1 | - | 5 | 659 | |||
AK164913.1 | - | 5 | 472 | |||
AK172045.1 | - | 5 | 646 | |||
AK178623.1 | - | 2 | 169 | |||
AK180908.1 | - | 2 | 169 | |||
AK198827.1 | - | 2 | 169 | |||
AK215101.1 | - | 2 | 169 | |||
AY255531.1 | - | 3 | 217 | |||
BC010244.1 | - | 3 | 271 | |||
BC028977.1 | - | 5 | 629 | 5363085 | ||
BC057296.1 | - | 5 | 648 |