Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011905.3 | NP_036035.3 | 3 | 2807 | ||
GenBank mRNA | AF124741.1 | - | 3 | 2746 | ||
AF165189.1 | - | 3 | 2771 | |||
AF216289.1 | - | 3 | 2770 | |||
AK005043.1 | - | 3 | 2808 | |||
AK154504.1 | - | 3 | 2777 | |||
BC014693.1 | - | 3 | 2966 | 3966249 |