Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vsnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012038.4 | NP_036168.1 | 2 | 444 | ||
GenBank mRNA | AJ489331.1 | - | 2 | 408 | ||
AK144454.1 | - | 2 | 374 | |||
AK160224.1 | - | 2 | 376 | |||
AY101375.1 | - | 1 | 146 | |||
BC046226.1 | - | 2 | 349 | 5692284 | ||
D21165.1 | - | 1 | 146 | |||
EU373446.1 | - | 1 | 146 |