Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013487.3 | NP_038515.3 | 2 | 198 | ||
GenBank mRNA | AK042172.1 | - | 2 | 200 | ||
AK050629.1 | - | 2 | 187 | |||
AK153843.1 | - | 2 | 163 | |||
AK154064.1 | - | 2 | 160 | |||
BC062807.1 | - | 2 | 103 | 1366111 | ||
EF382667.1 | - | 2 | 98 | |||
HM536173.1 | - | 2 | 98 | |||
X02339.1 | - | 2 | 66 |