Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081457.2 | NP_001074926.1 | 7 | 851 | ||
NM_001081458.2 | NP_001074927.1 | 7 | 851 | |||
NM_001135001.1 | NP_001128473.1 | 9 | 1082 | |||
NM_012023.3 | NP_036153.2 | 7 | 851 | |||
GenBank mRNA | AB055635.1 | - | 7 | 701 | ||
AB055636.1 | - | 7 | 701 | |||
AK031584.1 | - | 7 | 854 | |||
AK129042.1 | - | 9 | 954 | |||
AK191098.1 | - | 2 | 67 | |||
AK196453.1 | - | 2 | 67 | |||
BC003979.1 | - | 7 | 811 | 3489999 | ||
U37353.1 | - | 7 | 680 |