Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 2 | 554 | ||
NM_001317387.1 | NP_001304316.1 | 2 | 554 | |||
NM_001650.5 | NP_001641.1 | 2 | 554 | |||
NM_004028.4 | NP_004019.1 | 1 | 681 | |||
XM_011525942.2 | XP_011524244.1 | 2 | 596 | |||
GenBank mRNA | AB128929.1 | - | 3 | 84 | ||
AK222684.1 | - | 2 | 466 | |||
AK295069.1 | - | 3 | 365 | |||
BC022286.1 | - | 2 | 419 | 4717755 | ||
BI596912.1 | - | 2 | 454 | |||
BI667387.1 | - | 2 | 472 | |||
D63412.1 | - | 2 | 419 | |||
HQ901095.1 | - | 2 | 528 | |||
KF055862.1 | - | 2 | 399 | |||
U34845.1 | - | 1 | 617 | |||
U63622.1 | - | 2 | 438 | |||
U63623.1 | - | 1 | 342 |