Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001690.3 | NP_001681.2 | 3, 4 | 311 | ||
GenBank mRNA | AF113129.1 | - | 3, 4 | 238 | ||
AK293804.1 | - | 4, 5 | 345 | |||
AK314779.1 | - | 3, 4 | 274 | |||
AL514316.3 | - | 3, 4 | 281 | |||
BC013138.1 | - | 3, 4 | 259 | 4454722 | ||
BT006672.1 | - | 2, 3 | 203 | |||
DC350361.1 | - | 3, 4 | 311 |