Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arhgef28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012026.2 | NP_036156.2 | 31, 32 | 4177 | ||
GenBank mRNA | AK029167.1 | - | 25, 26 | 3337 | ||
BC098331.1 | - | 31, 32 | 4166 | |||
BC138030.1 | - | 31, 32 | 4101 | 8861049 | ||
U73199.1 | - | 31, 32 | 4177 |