Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013463.2 | NP_038491.2 | 7 | 2370 | ||
GenBank mRNA | AK040081.1 | - | 7 | 2302 | ||
AK164231.1 | - | 7 | 2303 | |||
U34071.1 | - | 7 | 2327 |