Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013499.2 | NP_038527.2 | 1 | 343 | ||
GenBank mRNA | AK004825.1 | - | 1 | 334 | ||
AK017702.1 | - | 1 | 307 | |||
AK134226.1 | - | 117 | ||||
BC028945.1 | - | 1 | 110 | 5371665 | ||
CA491482.1 | - | 380 | ||||
M23446.1 | - | 1 | 46 |