Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013606.1 | NP_038634.1 | 2, 3 | 211 | ||
GenBank mRNA | AB029920.1 | - | 2, 3 | 211 | ||
AK143831.1 | - | 2, 3 | 214 | |||
AK147949.1 | - | 833 | ||||
BC007127.1 | - | 2, 3 | 196 | 3591798 | ||
J03368.1 | - | 2, 3 | 211 |