Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113213.1 | NP_001106684.1 | 5 | 678 | ||
NM_001113214.1 | NP_001106685.1 | 5 | 678 | |||
NM_001177659.1 | NP_001171130.1 | 5 | 678 | |||
NM_001177661.1 | NP_001171132.1 | 4 | 524 | |||
NM_013615.3 | NP_038643.1 | 5 | 651 | |||
GenBank mRNA | AF000968.1 | - | 3 | 498 | ||
AF034105.1 | - | 5 | 614 | |||
AK143726.1 | - | 5 | 582 | |||
AK148523.1 | - | 5 | 591 | |||
AK165588.1 | - | 5 | 582 | |||
AK168415.1 | - | 5 | 580 | |||
BC057001.1 | - | 6 | 735 | 6827260 | ||
BQ840288.1 | - | 3 | 236 | |||
DQ091767.1 | - | 5 | 523 | |||
DQ091770.1 | - | 5 | 582 |