Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013552.2 | NP_038580.2 | 10 | 1072 | ||
GenBank mRNA | AF031932.1 | - | 10 | 1057 | ||
AF079222.1 | - | 11 | 1021 | |||
AK143123.1 | - | 10 | 1072 | |||
AK161734.1 | - | 1 | 2370 | |||
AK163410.1 | - | 10 | 1015 | |||
BC021427.1 | - | 10 | 1070 | 5035341 | ||
X64550.1 | - | 6 | 685 |