Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013654.3 | NP_038682.1 | 2 | 211 | ||
GenBank mRNA | AF128193.1 | - | 2 | 176 | ||
AF128194.1 | - | 2 | 176 | |||
AK078827.1 | - | 2 | 212 | |||
BC061126.1 | - | 2 | 209 | 30276350 | ||
BY163435.1 | - | 2 | 213 | |||
L04694.1 | - | 2 | 173 | |||
S50588.1 | - | 2 | 159 | |||
S71251.1 | - | 2 | 191 | |||
Z12297.1 | - | 2 | 159 |