Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000742.3 | NP_000733.2 | 7 | 2623 | ||
NM_001282455.1 | NP_001269384.1 | 7 | 2578 | |||
XM_005273397.2 | XP_005273454.1 | 7 | 2542 | |||
XM_006716282.1 | XP_006716345.1 | 8 | 2534 | |||
XM_011544388.1 | XP_011542690.1 | 8 | 2625 | |||
XM_011544389.1 | XP_011542691.1 | 7 | 2487 | |||
XM_017013003.1 | XP_016868492.1 | 7 | 2550 | |||
XM_017013004.1 | XP_016868493.1 | 8 | 2353 | |||
GenBank mRNA | AK308465.1 | - | 2 | 839 | ||
BC153866.1 | - | 6 | 2049 | 40121686 | ||
CD013901.1 | - | 6 | 2189 | |||
U62431.1 | - | 8 | 2209 |