Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000835.4 | NP_000826.2 | 6 | 1529 | ||
NM_001278553.1 | NP_001265482.1 | 6 | 1783 | |||
NR_103735.1 | 6 | 1529 | ||||
XM_006721845.3 | XP_006721908.1 | 5 | 2795 | |||
XM_006721846.3 | XP_006721909.1 | 5 | 2793 | |||
XM_011524686.2 | XP_011522988.1 | 5 | 2797 | |||
XM_011524687.2 | XP_011522989.1 | 5 | 2797 | |||
XM_011524688.2 | XP_011522990.1 | 5 | 2796 | |||
XM_011524689.2 | XP_011522991.1 | 6 | 1520 | |||
XM_011524692.2 | XP_011522994.1 | 5 | 2793 | |||
XM_017024544.1 | XP_016880033.1 | 5 | 2798 | |||
GenBank mRNA | AB208799.1 | - | 3 | 4732 | ||
BC041128.1 | - | 6 | 1475 | |||
BC140801.1 | - | 6 | 1407 | 9021669 | ||
L76224.1 | - | 5 | 1373 | |||
U77782.1 | - | 6 | 1561 |