Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013721.3 | NP_038749.1 | 4, 5 | 444 | ||
GenBank mRNA | AK019449.1 | - | 4, 5 | 447 | ||
AK135447.1 | - | 4, 5 | 450 | |||
AK160044.1 | - | 4, 5 | 446 | |||
AK160621.1 | - | 4, 5 | 445 | |||
AK160622.1 | - | 4, 5 | 452 | |||
BC016489.1 | - | 1, 2 | 65 | |||
BC029992.1 | - | 3, 4 | 433 | |||
BC052339.1 | - | 3, 4 | 437 | |||
BC065176.1 | - | 3, 4 | 432 | 6819288 | ||
BC080663.1 | - | 3, 4 | 433 | 4487611 | ||
BC080669.1 | - | 3, 4 | 433 | 5060194 | ||
BC080712.1 | - | 3, 4 | 428 | 4457484 | ||
BC084678.1 | - | 3, 4 | 445 | 30431224 | ||
BC089624.1 | - | 4, 5 | 449 | 6763528 | ||
BC091731.1 | - | 3, 4 | 428 | 2655142 | ||
BC091769.1 | - | 3, 4 | 433 | 4158894 | ||
BC099389.1 | - | 3, 4 | 428 | 30526878 |
Gene Symbol: Gm5766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030194.1 | - | 1 | 1494 | ||
BC147642.1 | - | 1 | 856 |