Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mixl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013729.3 | NP_038757.1 | 2 | 567 | ||
GenBank mRNA | AB221547.1 | - | 2 | 549 | ||
AF135063.1 | - | 2 | 545 | |||
AF154573.1 | - | 2 | 567 | |||
AF201959.1 | - | 2 | 510 | |||
BC099876.1 | - | 2 | 567 | 40040047 | ||
BC099877.1 | - | 2 | 567 | 40040048 |