Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048146.2 | NP_001041611.1 | 2 | 609 | ||
NM_001286507.1 | NP_001273436.1 | 2 | 609 | |||
NM_001286508.1 | NP_001273437.1 | 2 | 527 | |||
NM_013727.4 | NP_038755.1 | 2 | 609 | |||
GenBank mRNA | AB007141.1 | - | 2 | 445 | ||
AK004992.1 | - | 2 | 588 | |||
AK015942.1 | - | 2 | 430 | |||
AK030076.1 | - | 2 | 441 | |||
AK036102.1 | - | 2 | 516 | |||
AK134714.1 | - | 2 | 512 | |||
AK153477.1 | - | 2 | 573 | |||
AK165549.1 | - | 2 | 495 | |||
AK167783.1 | - | 2 | 553 | |||
BC006851.1 | - | 2 | 504 | 3156857 |