Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285.3 | NP_001276.2 | 5 | 839 | ||
GenBank mRNA | AF039400.1 | - | 6 | 1061 | ||
AF127036.1 | - | 5 | 714 | |||
AK291999.1 | - | 5 | 755 | |||
AK300858.1 | - | 1 | 125 |