Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013763.2 | NP_038791.2 | 4 | 566 | ||
GenBank mRNA | AF097523.1 | - | 4 | 553 | ||
AK010899.1 | - | 4 | 586 | |||
AK132366.1 | - | 4 | 592 | |||
AK165686.1 | - | 4 | 586 | |||
BC039273.1 | - | 4 | 599 | 5051833 | ||
BC048185.1 | - | 4 | 566 | 6827496 |