Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013781.3 | NP_038809.1 | 2 | 373 | ||
GenBank mRNA | AB043953.1 | - | 2 | 356 | ||
AF168364.1 | - | 2 | 314 | |||
AK155165.1 | - | 2 | 309 | |||
AK172072.1 | - | 2 | 294 | |||
BC113203.1 | - | 2 | 328 | 40048064 |