Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113374.1 | NP_001106845.1 | 3 | 262 | ||
NM_001113375.1 | NP_001106846.1 | 3 | 262 | |||
NM_013826.3 | NP_038854.2 | 4 | 384 | |||
GenBank mRNA | AF091872.1 | - | 4 | 335 | ||
AK002719.1 | - | 3 | 230 | |||
AK010112.1 | - | 4 | 364 | |||
AK078782.1 | - | 3 | 230 | |||
AK205528.1 | - | 2 | 68 | |||
AK214585.1 | - | 2 | 68 | |||
BC004631.1 | - | 3 | 234 | |||
BC024371.1 | - | 4 | 344 | 4983231 | ||
CX220128.1 | - | 3 | 212 |