Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013822.5 | NP_038850.1 | 26 | 3682 | ||
GenBank mRNA | AF171092.1 | - | 26 | 3687 | ||
AK078018.1 | - | 3 | 448 | |||
AK137092.1 | - | 26 | 3582 | |||
AK162228.1 | - | 9 | 1184 | |||
BC058675.1 | - | 26 | 3494 | 6834418 |