Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013907.2 | NP_038935.1 | 2 | 434 | ||
GenBank mRNA | AB022163.1 | - | 2 | 434 | ||
AF176519.1 | - | 2 | 412 | |||
BC027031.1 | - | 2 | 296 | 5364017 |