Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272035.1 | NP_001258964.1 | 1 | 878 | ||
NM_015728.5 | NP_056543.2 | 1 | 878 | |||
GenBank mRNA | AB016795.1 | - | 1 | 335 | ||
AK135092.1 | - | 1 | 807 | |||
AK136323.1 | - | 1 | 873 | |||
AK144703.1 | - | 1 | 744 | |||
BC005616.1 | - | 1 | 455 | 3154886 | ||
BC054395.1 | - | 1 | 878 | 6492236 | ||
BC063104.1 | - | 811 | 5042025 |