Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177730.1 | NP_001171201.1 | 4, 5 | 686 | ||
NM_013839.4 | NP_038867.2 | 4, 5 | 839 | |||
GenBank mRNA | AF085745.1 | - | 4, 5 | 581 | ||
AJ132601.1 | - | 4, 5 | 617 | |||
AK143904.1 | - | 4, 5 | 620 | |||
AK159250.1 | - | 4, 5 | 840 | |||
AY195870.1 | - | 4, 5 | 686 | |||
AY195871.1 | - | 4, 5 | 686 | |||
BC012646.1 | - | 4, 5 | 627 | 4163109 |