Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013852.2 | NP_038880.1 | 5 | 453 | ||
GenBank mRNA | AK030626.1 | - | 5 | 417 | ||
AK183731.1 | - | 3 | 155 | |||
AK189060.1 | - | 3 | 154 | |||
AK214630.1 | - | 2 | 84 | |||
BC032923.1 | - | 5 | 394 | 1430389 | ||
CD540120.1 | - | 5 | 453 |