Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013867.2 | NP_038895.1 | 12 | 2992 | ||
GenBank mRNA | AF179566.1 | - | 12 | 2940 | ||
AK143894.1 | - | 12 | 2780 | |||
AK167578.1 | - | 12 | 2810 | |||
BC023930.1 | - | 12 | 2981 | 5324365 |