Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163447.1 | NP_001156919.1 | 2 | 548 | ||
NM_001163448.1 | NP_001156920.1 | 2 | 548 | |||
NM_001163449.1 | NP_001156921.1 | 2 | 548 | |||
NM_001163450.1 | NP_001156922.1 | 2 | 548 | |||
NM_001163451.1 | NP_001156923.1 | 2 | 548 | |||
NM_001163453.1 | NP_001156925.1 | 2 | 548 | |||
NM_013931.4 | NP_038959.2 | 2 | 548 | |||
GenBank mRNA | AB005662.1 | - | 2 | 531 | ||
AB043123.1 | - | 2 | 425 | |||
AB043124.1 | - | 2 | 425 | |||
AB043125.1 | - | 2 | 425 | |||
AB093282.2 | - | 2 | 500 | |||
AF178636.1 | - | 2 | 465 | |||
AF178637.1 | - | 2 | 541 | |||
AF262046.1 | - | 2 | 531 | |||
AK163208.1 | - | 2 | 517 | |||
BC060603.1 | - | 2 | 483 | 5706805 | ||
BC065105.1 | - | 2 | 538 | 30356817 | ||
BC068312.1 | - | 2 | 526 | 30100914 |