Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272035.1 | NP_001258964.1 | 4 | 1764 | ||
NM_015728.5 | NP_056543.2 | 4 | 1764 | |||
GenBank mRNA | AB016795.1 | - | 4 | 1221 | ||
AK144703.1 | - | 4 | 1631 | |||
AW519598.1 | - | 2 | 243 | |||
BC005616.1 | - | 4 | 1341 | 3154886 | ||
BC054395.1 | - | 4 | 1764 | 6492236 | ||
BC063104.1 | - | 1697 | 5042025 |