Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015821.2 | NP_056636.2 | 3 | 664 | ||
GenBank mRNA | AF176523.1 | - | 3 | 589 | ||
AK053206.1 | - | 4 | 832 | |||
AK155194.1 | - | 3 | 625 | |||
AK170367.1 | - | 3 | 624 | |||
BC023902.1 | - | 2 | 529 | 5321302 | ||
CX222518.1 | - | 3 | 632 |