Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC541473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310313.1 | - | 4 | 553 | ||
HY019943.1 | - | 3 | 425 |
Gene Symbol: FKBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135211.2 | NP_001128683.1 | 4 | 440 | ||
NM_001281304.1 | NP_001268233.1 | 3 | 441 | |||
NM_003602.4 | NP_003593.3 | 4 | 531 | |||
XM_006716153.2 | XP_006716216.1 | 4 | 435 | |||
XM_017012741.1 | XP_016868230.1 | 4 | 435 | |||
XM_017012742.1 | XP_016868231.1 | 4 | 438 | |||
GenBank mRNA | AF038847.1 | - | 4 | 395 | ||
AF447060.1 | - | 3 | 398 | |||
AK302121.1 | - | 4 | 428 | |||
AK310255.1 | - | 4 | 431 | |||
BC036817.1 | - | 4 | 403 | 5744865 | ||
BI461016.1 | - | 4 | 444 | |||
DB033483.1 | - | 4 | 466 |