Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077202.2 | NP_001070670.1 | 2, 3 | 955 | ||
NM_001290468.1 | NP_001277397.1 | 1, 2 | 596 | |||
GenBank mRNA | AB024565.1 | - | 1, 2 | 518 | ||
AK043774.1 | - | 1, 2 | 79 | |||
AK052348.1 | - | 2, 3 | 735 | |||
AK083281.1 | - | 1, 2 | 583 |