Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015821.2 | NP_056636.2 | 3 | 1869 | ||
GenBank mRNA | AF176523.1 | - | 3 | 1794 | ||
AK155194.1 | - | 3 | 1830 | |||
AK170367.1 | - | 3 | 1829 | |||
AK181171.1 | - | 1 | 352 | |||
AK188074.1 | - | 1 | 287 | |||
BC023902.1 | - | 2 | 1734 | 5321302 |