Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016676.5 | NP_057885.1 | 6 | 3189 | ||
GenBank mRNA | AK012933.1 | - | 1 | 316 | ||
AK028320.1 | - | 6 | 3160 | |||
AK087964.1 | - | 6 | 3151 | |||
AK146357.1 | - | 6 | 3190 | |||
AK150319.1 | - | 6 | 3149 | |||
AK150384.1 | - | 6 | 2843 | |||
AK150791.1 | - | 6 | 3140 | |||
AK151473.1 | - | 6 | 2843 | |||
AK153315.1 | - | 6 | 3149 | |||
AK153325.1 | - | 6 | 3139 | |||
AK159383.1 | - | 6 | 3155 | |||
AK159845.1 | - | 6 | 3155 | |||
AK159988.1 | - | 6 | 3149 | |||
AK160960.1 | - | 6 | 3144 | |||
AK166010.1 | - | 6 | 3163 | |||
AK168320.1 | - | 6 | 3149 | |||
BC052735.1 | - | 3273 | ||||
BC056374.1 | - | 6 | 3140 | 6848840 |