Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016699.2 | NP_057908.2 | 2 | 259 | ||
GenBank mRNA | AF091392.1 | - | 2 | 228 | ||
AK017621.1 | - | 2 | 232 | |||
AK081014.1 | - | 2 | 255 | |||
AK144162.1 | - | 2 | 244 | |||
AK162887.1 | - | 2 | 228 | |||
AK166070.1 | - | 2 | 243 | |||
AK168700.1 | - | 2 | 232 | |||
AK199580.1 | - | 1 | 69 | |||
BC027326.1 | - | 2 | 237 | 3987812 | ||
BC027840.1 | - | 2 | 247 | 5325071 | ||
BY277385.1 | - | 2 | 267 |