Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038710.1 | NP_001033799.1 | 2 | 522 | ||
NM_016711.3 | NP_057920.2 | 2 | 458 | |||
GenBank mRNA | AF177170.1 | - | 2 | 407 | ||
AF237629.1 | - | 3 | 366 | |||
AK015263.1 | - | 2 | 503 | |||
AK017072.1 | - | 2 | 503 | |||
AK018223.1 | - | 2 | 255 | |||
AK076562.1 | - | 2 | 472 | |||
AK079077.1 | - | 2 | 302 | |||
AK147581.1 | - | 2 | 454 | |||
AK164178.1 | - | 2 | 304 | |||
BC052227.1 | - | 2 | 522 | |||
BC061124.1 | - | 2 | 288 | 6531518 |