Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABCG2 SIRNA Assays ›
Gene Symbol:
ABCG2
Gene Name:
ATP binding cassette subfamily G member 2 (Junior blood group)
Gene Aliases:
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1
Chromosome Location:
Chr.4: 88090264 - 88231417 on Build GRCh38
UniGene ID:
Hs.480218
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7244
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABCG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257386.1
NP_001244315.1
2
443
NM_004827.2
NP_004818.2
2
588
XM_005263354.3
XP_005263411.1
2
431
XM_005263355.3
XP_005263412.1
2
538
XM_005263356.3
XP_005263413.1
2
314
XM_011532420.2
XP_011530722.1
4
702
XM_017008852.1
XP_016864341.1
2
431
XM_017008853.1
XP_016864342.1
2
228
GenBank mRNA
AB051855.1
-
2
139
AB056867.1
-
1
95
AF098951.2
-
2
333
AF103796.1
-
2
299
AF463519.1
-
1
103
AK290000.1
-
2
159
AY017168.1
-
2
288
AY289766.1
-
2
295
AY333755.1
-
1
104
AY333756.1
-
1
104
BC021281.2
-
2
588
4908836
BC092408.1
-
2
443
6181905
BI598784.1
-
2
646
DQ459561.1
-
2
217
DQ459562.1
-
2
217
DQ459563.1
-
2
217
DQ459565.1
-
2
208
DQ459566.1
-
2
210
DQ459568.1
-
2
209
DQ459570.1
-
2
300
DQ459571.1
-
2
300
DQ459573.1
-
2
190
DQ459574.1
-
2
149
DQ459575.1
-
2
143
DQ459576.1
-
2
215
DQ459577.1
-
2
213
DQ459578.1
-
2
209
DQ459579.1
-
2
209
DQ459580.1
-
2
166
DQ459581.1
-
2
161
DQ459582.1
-
2
400
DQ459583.1
-
2
352
DQ459584.1
-
2
352
DQ459585.1
-
2
351
DQ459586.1
-
2
349
DQ459587.1
-
2
342
DQ459588.1
-
2
245
DQ459589.1
-
2
183
DQ459591.1
-
2
205
DQ459592.1
-
2
193
DQ459593.1
-
2
233
DQ459594.1
-
2
233
DQ459595.1
-
2
210
DQ459596.1
-
2
208
DQ459597.1
-
2
328
DQ459598.1
-
2
300
DQ459599.1
-
2
300
DQ459600.1
-
2
193
DQ459601.1
-
2
193
DQ459603.1
-
2
146
DQ459604.1
-
2
209
DQ459605.1
-
2
173
DQ459606.1
-
2
165
DQ459607.1
-
1
109
DQ459608.1
-
1
34
DQ459610.1
-
2
195
DQ459611.1
-
2
193
DQ459612.1
-
2
189
DQ459613.1
-
2
189
HQ386990.1
-
2
193
HQ386991.1
-
2
193
HQ386992.1
-
2
193
HQ386993.1
-
2
193
HQ386994.1
-
2
193
HQ386995.1
-
2
193
HQ386996.1
-
2
193
HQ386997.1
-
2
193
HQ386998.1
-
2
352
HQ386999.1
-
2
352
HQ387000.1
-
2
352
HQ387001.1
-
2
352
HQ387002.1
-
2
352
HQ387003.1
-
2
352
HQ387004.1
-
2
352
HQ387005.1
-
2
352
HQ387006.1
-
2
400
HQ387007.1
-
2
209
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
Gene Ontology Categories:
Function(s)
Process(es)
transport
drug transmembrane transport
cellular iron ion homeostasis
response to iron ion
heme transport
response to drug
xenobiotic transport
urate metabolic process
drug export
response to folic acid
embryonic process involved in female pregnancy
cellular response to dexamethasone stimulus
transporter activity
protein binding
ATP binding
xenobiotic-transporting ATPase activity
heme transporter activity
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0