Search Thermo Fisher Scientific
Gene Symbol: Sfrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016687.3 | NP_057896.1 | 2 | 757 | ||
GenBank mRNA | AF117709.1 | - | 2 | 772 | ||
AK080766.1 | - | 2 | 758 | |||
BC034853.1 | - | 2 | 716 | 1364770 |
Set Membership: |
![]() |