Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004975.3 | NP_004966.1 | 1 | 650 | ||
XM_006723784.3 | XP_006723847.1 | 2 | 744 | |||
XM_011528799.2 | XP_011527101.1 | 2 | 773 | |||
GenBank mRNA | AF026005.1 | - | 1 | 486 | ||
BC110572.1 | - | 2 | 351 | |||
BC110573.1 | - | 2 | 351 | |||
L02840.1 | - | 1 | 672 |