Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016690.4 | NP_057899.2 | 2 | 528 | ||
GenBank mRNA | AB017020.1 | - | 2 | 234 | ||
AK011581.1 | - | 2 | 217 | |||
AK011767.1 | - | 2 | 217 | |||
AK172629.1 | - | 2 | 187 | |||
BC021374.1 | - | 2 | 186 | 5039077 | ||
BM950445.1 | - | 2 | 528 |