Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289653.1 | NP_001276582.1 | 4, 5 | 396 | ||
GenBank mRNA | AK035019.1 | - | 2, 3 | 249 | ||
AK039790.1 | - | 460 | ||||
AK048913.1 | - | 667 | ||||
BC051968.1 | - | 3, 4 | 364 | 6401465 | ||
BU612839.1 | - | 4, 5 | 605 | |||
U59230.1 | - | 4, 5 | 397 |