Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130479.2 | NP_001123951.1 | 5 | 618 | ||
GenBank mRNA | AJ222586.1 | - | 5 | 618 | ||
AK145934.1 | - | 5 | 573 | |||
AK160442.1 | - | 4 | 466 | |||
BC010459.1 | - | 5 | 532 | 3585076 |